missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyl Isocyanate 98.0+%, TCI America™

Catalog No. i07665g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07665G 5 g
I076625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07665G Supplier TCI America Supplier No. I07665G

Chemical Identifiers

CAS 16588-74-2
Molecular Formula C9H3F6NO
Molecular Weight (g/mol) 255.119
MDL Number MFCD00013559
InChI Key NRSSOFNMWSJECS-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylisocyanate, 3,5-bis trifluoromethyl phenyl isocyanate, 1-isocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl isocyanate, 3,5-di trifluoromethyl phenyl isocyanate, 3,5-di trifluoromethyl phenylisocyanate, 3,5-bis trifluoromethyl benzenisocyanate, pubchem2735, acmc-209dtj, 3,5-bistrifluorometylphenylisocyanate
PubChem CID 2733314
IUPAC Name 1-isocyanato-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)N=C=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 78°C
Quantity 5 g
UN Number 2206
Formula Weight 255.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyl Isocyanate