missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%

Catalog No. p-70495
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314170010 1 g Glass bottle
AC314170050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314170010 Supplier Thermo Scientific Chemicals Supplier No. 314170010

CAS: 16588-74-2 | C9H3F6NO | 255.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16588-74-2
Molecular Formula C9H3F6NO
Molecular Weight (g/mol) 255.12
MDL Number MFCD00013559
InChI Key NRSSOFNMWSJECS-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylisocyanate, 3,5-bis trifluoromethyl phenyl isocyanate, 1-isocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl isocyanate, 3,5-di trifluoromethyl phenyl isocyanate, 3,5-di trifluoromethyl phenylisocyanate, 3,5-bis trifluoromethyl benzenisocyanate, pubchem2735, acmc-209dtj, 3,5-bistrifluorometylphenylisocyanate
PubChem CID 2733314
IUPAC Name 1-isocyanato-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)N=C=O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.4700g/mL
Boiling Point 62.0°C (2.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4295 to 1.4315
Linear Formula (CF3)2C6H3NCO
Quantity 1 g
Specific Gravity 1.47
Formula Weight 255.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)phenyl isocyanate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
TSCA TSCA

RUO – Research Use Only