missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenol 96.0+%, TCI America™

Catalog No. p-7119320
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18695G 5 g
B186925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18695G Supplier TCI America Supplier No. B18695G
Only null left

Chemical Identifiers

CAS 349-58-6
Molecular Formula C8H4F6O
Molecular Weight (g/mol) 230.109
MDL Number MFCD00000386
InChI Key ODSXJQYJADZFJX-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
PubChem CID 67680
IUPAC Name 3,5-bis(trifluoromethyl)phenol
SMILES C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
Formula Weight 230.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenol