missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzylamine, 95%

Catalog No. AC305640010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305640010 1 g Glass bottle
AC305640050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305640010 Supplier Thermo Scientific Chemicals Supplier No. 305640010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85068-29-7 | C9H7F6N | 243.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85068-29-7
Molecular Formula C9H7F6N
Molecular Weight (g/mol) 243.15
MDL Number MFCD00009909
InChI Key DHVHORCFFOSRBP-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
PubChem CID 521099
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]methanamine
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 46°C to 55°C
Color White to Beige
Boiling Point 82°C to 84°C (15.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CH2NH2
Quantity 1 g
Formula Weight 243.15
Percent Purity 95%
Physical Form Crystalline Mass, Chunks or Crystals and Powder
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 285-290-9

RUO – Research Use Only