missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™

Catalog No. B17525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17525G Supplier TCI America Supplier No. B17525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32707-89-4
Molecular Formula C9H6F6O
Molecular Weight (g/mol) 244.136
MDL Number MFCD00009907
InChI Key BJTWPJOGDWRYDD-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl alcohol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl-phenyl-methanol, 3,5-ditrifluoromethylbenzyl alcohol, benzenemethanol, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylalcohol, 3,5-di trifluoromethyl benzyl alcohol, 3,5-di trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl benzyl alcohol
PubChem CID 122933
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]methanol
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CO

Specifications

Melting Point 55°C
Color White
Quantity 5 g
Formula Weight 244.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl Alcohol