missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(trifluoromethyl)benzonitrile 98.0+%, TCI America™
Supplier: TCI America B189625G
Specifications
| 3,5-Bis(trifluoromethyl)benzonitrile | |
| White | |
| MFCD00000379 | |
| 3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene | |
| FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F | |
| 239.12 | |
| 239.12 | |
| Crystalline Lumps |
| 27126-93-8 | |
| C9H3F6N | |
| 25 g | |
| CZKHHAOIHXHOSR-UHFFFAOYSA-N | |
| 3,5-bis(trifluoromethyl)benzonitrile | |
| 96746 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 27126-93-8 | |
| 239.12 | |
| CZKHHAOIHXHOSR-UHFFFAOYSA-N | |
| 96746 | |
| FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F |
| C9H3F6N | |
| MFCD00000379 | |
| 3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene | |
| 3,5-bis(trifluoromethyl)benzonitrile |
Safety and Handling
TSCA : No