missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. B18965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18965G 5 g
B189625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18965G Supplier TCI America Supplier No. B18965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27126-93-8
Molecular Formula C9H3F6N
Molecular Weight (g/mol) 239.12
MDL Number MFCD00000379
InChI Key CZKHHAOIHXHOSR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene
PubChem CID 96746
IUPAC Name 3,5-bis(trifluoromethyl)benzonitrile
SMILES FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F

Specifications

Color White
Quantity 5 g
Formula Weight 239.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzonitrile