missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzoic acid, 98%

Catalog No. AAA1132506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1132506 5 g
AAA1132514 25 g
AAA1132522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1132506 Supplier Thermo Scientific Chemicals Supplier No. A1132506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 725-89-3 | C9H4F6O2 | 258.12 g/mol

In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In vitro metabolism studies on a series of 3,5-bis(trifluoromethyl)benzyl ethers have identified 3,5-bis(trifluoromethyl)benzoic acid as a significant metabolite possibly arising via oxidation of the benzylic position. The reactivity of the hydroxy- propyl units was demonstrated by the esterification of por- phyrazine derivatives with 3,5-bis(trifluoromethyl)benzoic acid in the presence of dicyclohexylcarbodiimide and toluene- p-sulfonic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 725-89-3
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.12
MDL Number MFCD00000388
InChI Key HVFQJWGYVXKLTE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
PubChem CID 12889
IUPAC Name 3,5-bis(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 140°C to 144°C
Density 1.42
Quantity 5 g
Beilstein 2058600
Solubility Information Slightly soluble in water.
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 3,5-Bis (trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-970-1
RTECSNumber DG4448020
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only