missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzhydrazide, 97%

CAS: 26107-82-4 | C9H6F6N2O | 272.15 g/mol

Supplier:  Thermo Scientific Chemicals A1349214

 View more versions of this product

Catalog No. AAA1349214


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Bis(trifluoromethyl)benzhydrazide
132°C to 134°C
MFCD00051848
2596448
GBBRFBSFWKFTMZ-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzohydrazide
100265
97%
26107-82-4
C9H6F6N2O
25 g
3,5-bis trifluoromethyl benzhydrazide, 3,5-bis trifluoromethyl benzohydrazide, 3,5-di trifluoromethyl benzhydrazide, 3,5-bis-trifluoromethylbenzoic acid hydrazide, 3,5-bis trifluoromethyl benzoic acid hydrazide, benzoic acid, 3,5-bis trifluoromethyl-, hydrazide, 3,5-bis-trifluoromethyl-benzhydrazide, maybridge1_001057, acmc-1cp6z, ksc206c6d
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)NN
272.15
272.15

Chemical Identifiers

26107-82-4
272.15
GBBRFBSFWKFTMZ-UHFFFAOYSA-N
100265
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)NN
C9H6F6N2O
MFCD00051848
3,5-bis trifluoromethyl benzhydrazide, 3,5-bis trifluoromethyl benzohydrazide, 3,5-di trifluoromethyl benzhydrazide, 3,5-bis-trifluoromethylbenzoic acid hydrazide, 3,5-bis trifluoromethyl benzoic acid hydrazide, benzoic acid, 3,5-bis trifluoromethyl-, hydrazide, 3,5-bis-trifluoromethyl-benzhydrazide, maybridge1_001057, acmc-1cp6z, ksc206c6d
3,5-bis(trifluoromethyl)benzohydrazide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 247-462-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only