missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzhydrazide, 97%

Catalog No. AAA1349203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1349203 1 g
AAA1349206 5 g
AAA1349214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1349203 Supplier Thermo Scientific Chemicals Supplier No. A1349203

CAS: 26107-82-4 | C9H6F6N2O | 272.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26107-82-4
Molecular Formula C9H6F6N2O
Molecular Weight (g/mol) 272.15
MDL Number MFCD00051848
InChI Key GBBRFBSFWKFTMZ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzhydrazide, 3,5-bis trifluoromethyl benzohydrazide, 3,5-di trifluoromethyl benzhydrazide, 3,5-bis-trifluoromethylbenzoic acid hydrazide, 3,5-bis trifluoromethyl benzoic acid hydrazide, benzoic acid, 3,5-bis trifluoromethyl-, hydrazide, 3,5-bis-trifluoromethyl-benzhydrazide, maybridge1_001057, acmc-1cp6z, ksc206c6d
PubChem CID 100265
IUPAC Name 3,5-bis(trifluoromethyl)benzohydrazide
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)NN

Specifications

Melting Point 132°C to 134°C
Quantity 1 g
Beilstein 2596448
Formula Weight 272.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-462-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only