missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™
$80.85 - $238.72
Chemical Identifiers
| CAS | 39234-86-1 |
|---|---|
| Molecular Formula | C8H3ClF6O2S |
| Molecular Weight (g/mol) | 312.61 |
| MDL Number | MFCD00014725 |
| InChI Key | BTRCVKADYDVSLI-UHFFFAOYSA-N |
| Synonym | 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724 |
| PubChem CID | 520945 |
| IUPAC Name | 3,5-bis(trifluoromethyl)benzenesulfonyl chloride |
| SMILES | C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F |
Chemical Identifiers
| 39234-86-1 | |
| 312.61 | |
| BTRCVKADYDVSLI-UHFFFAOYSA-N | |
| 520945 | |
| C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F |
| C8H3ClF6O2S | |
| MFCD00014725 | |
| 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724 | |
| 3,5-bis(trifluoromethyl)benzenesulfonyl chloride |
Specifications
| 39234-86-1 | |
| C8H3ClF6O2S | |
| 1 g | |
| 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724 | |
| C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F | |
| 312.61 | |
| 312.61 | |
| Crystals |
| White-Yellow | |
| MFCD00014725 | |
| 3261 | |
| BTRCVKADYDVSLI-UHFFFAOYSA-N | |
| 3,5-bis(trifluoromethyl)benzenesulfonyl chloride | |
| 520945 | |
| ≥98.0% (GC,T) | |
| 3,5-Bis(trifluoromethyl)benzenesulfonyl Chloride |
Safety and Handling
TSCA : No