missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. B21861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21861G 1 g
B21865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21861G Supplier TCI America Supplier No. B21861G

Chemical Identifiers

CAS 39234-86-1
Molecular Formula C8H3ClF6O2S
Molecular Weight (g/mol) 312.61
MDL Number MFCD00014725
InChI Key BTRCVKADYDVSLI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
PubChem CID 520945
IUPAC Name 3,5-bis(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 312.61
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzenesulfonyl Chloride