missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzaldehyde, 97%

Catalog No. AC307690250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307690250 25 g Glass bottle
AC307690010 1 g Glass bottle
AC307690050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC307690250 Supplier Thermo Scientific Chemicals Supplier No. 307690250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 401-95-6 | C9H4F6O | 242.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401-95-6
Molecular Formula C9H4F6O
Molecular Weight (g/mol) 242.12
MDL Number MFCD00010206
InChI Key LDWLIXZSDPXYDR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
PubChem CID 136247
IUPAC Name 3,5-bis(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 37°C (1.3 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4215 to 1.4235
Linear Formula (CF3)2C6H3CHO
Quantity 25 g
Specific Gravity 1.47
Formula Weight 242.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only