missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)aniline 97.0+%, TCI America™

Catalog No. B132425G
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B132425G 25 g
B132410G 10 g
B1324250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B132425G Supplier TCI America Supplier No. B132425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328-74-5
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.125
MDL Number MFCD00000394
InChI Key CDIDGWDGQGVCIB-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl
PubChem CID 9480
IUPAC Name 3,5-bis(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
Formula Weight 229.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)aniline
RTECSNumber ZE9800000
TSCA Yes