missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Bis(trifluoromethyl)acetophenone 98.0+%, TCI America™

Catalog No. p-7119374
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B19495G 5 g
B194925G 25 g
2 options

Catalog No. B19495G

Supplier: TCI America B19495G

Only null left

Chemical Identifiers

CAS 30071-93-3
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.15
MDL Number MFCD00009910
InChI Key MCYCSIKSZLARBD-UHFFFAOYSA-N
Synonym 3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
PubChem CID 121616
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-one
SMILES CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 256.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′,5′-Bis(trifluoromethyl)acetophenone