missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Bis(trifluoromethyl)acetophenone, 98%

CAS: 30071-93-3 | C10H6F6O | 256.15 g/mol

$96.74 - $323.05

Chemical Identifiers

CAS 30071-93-3
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.15
MDL Number MFCD00009910
InChI Key MCYCSIKSZLARBD-UHFFFAOYSA-N
Synonym 3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
PubChem CID 121616
SMILES CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1034106
View Documents
Thermo Scientific Chemicals
A1034106
5 g
Each for $96.74
Only null left
 
AAA1034114
View Documents
Thermo Scientific Chemicals
A1034114
25 g
Each for $323.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

30071-93-3
256.15
MCYCSIKSZLARBD-UHFFFAOYSA-N
121616
C10H6F6O
MFCD00009910
3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

30071-93-3
85°C (8 mmHg)
C10H6F6O
MFCD00009910
2384789
MCYCSIKSZLARBD-UHFFFAOYSA-N
1-[3,5-bis(trifluoromethyl)phenyl]ethanone
121616
98%
1.422
82°C (179°F)
1.423
5 g
3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
256.15
256.15
3',5'-Bis (trifluoromethyl)acetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

EINECSNumber : 250-023-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only