missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Bis(trifluoromethyl)acetophenone, 98%

Catalog No. AAA1034106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1034106 5 g
AAA1034114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1034106 Supplier Thermo Scientific Chemicals Supplier No. A1034106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 30071-93-3 | C10H6F6O | 256.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30071-93-3
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.15
MDL Number MFCD00009910
InChI Key MCYCSIKSZLARBD-UHFFFAOYSA-N
Synonym 3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
PubChem CID 121616
SMILES CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.422
Boiling Point 85°C (8 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.423
Quantity 5 g
Beilstein 2384789
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]ethanone
Formula Weight 256.15
Percent Purity 98%
Chemical Name or Material 3',5'-Bis (trifluoromethyl)acetophenone
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 250-023-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only