Learn More
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester, 97%
CAS: 944392-68-1 | C16H21BO6 | 320.148 g/mol
$331.88 - $983.27
Chemical Identifiers
| CAS | 944392-68-1 |
|---|---|
| Molecular Formula | C16H21BO6 |
| Molecular Weight (g/mol) | 320.148 |
| MDL Number | MFCD11858596 |
| InChI Key | IGSNWXAGFXHYOG-UHFFFAOYSA-N |
| PubChem CID | 42614529 |
| IUPAC Name | dimethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-dicarboxylate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)C(=O)OC)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3652503
|
Thermo Scientific Chemicals
H3652503 |
1 g |
Each for $331.88
|
|
|||||
|
AAH3652506
|
Thermo Scientific Chemicals
H3652506 |
5 g |
Each for $983.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 944392-68-1 | |
| 320.148 | |
| IGSNWXAGFXHYOG-UHFFFAOYSA-N | |
| dimethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-dicarboxylate |
| C16H21BO6 | |
| MFCD11858596 | |
| 42614529 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)C(=O)OC)C(=O)OC |
Specifications
| 944392-68-1 | |
| Pink | |
| MFCD11858596 | |
| dimethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isophthalate, 3,5-bis methoxycarbonyl phenylboronic acid pinacol ester, 3,5-bis methoxycarbonyl benzeneboronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,3-benzenedicarboxylic acid,1,3-dimethyl ester, 1,3-dimethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,3-dicarboxylate, 3,5-dimethyl benzenecarboxylate-5-boronic acid pinacol ester, dimethyl 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl isophthalate, dimethyl5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isophthalate, 1,3-dimethyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene-1,3-dicarboxylate, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isophthalic acid dimethyl ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)C(=O)OC)C(=O)OC | |
| 320.148 | |
| 320.15 | |
| Solid |
| 128°C | |
| C16H21BO6 | |
| 1 g | |
| IGSNWXAGFXHYOG-UHFFFAOYSA-N | |
| dimethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-dicarboxylate | |
| 42614529 | |
| 97% | |
| 3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only