missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(4-nitrophenoxy)benzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 232°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 396.31 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,5-Bis(4-nitrophenoxy)benzoic Acid |
Safety and Handling
| TSCA | No |