missing translation for 'onlineSavingsMsg'
Learn More

3,5,7-Trimethyladamantane-1-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-2699425
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB12973CB 250 mg Amber Glass Bottle
BTB12973DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB12973CB Supplier Thermo Scientific Chemicals Supplier No. BTB12973CB

Chemical Identifiers

CAS 15291-66-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00187581
InChI Key DHIIUPVOLLOORM-UHFFFAOYSA-N
Synonym 3,5,8-trimethyladamantanecarboxylic acid, maybridge4_000576, 1,3,5-trimethyladamantane-7-carboxylic acid, 3,5,7-trimethyl adamantane-1-carboxylic acid, 1s,3r,5s,7s-3,5,7-trimethyladamantane-1-carboxylic acid, 3r,5s,7s-3,5,7-trimethyladamantane-1-carboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylicacid, 3,5,7-trimethyl, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3,5,7-trimethyl
PubChem CID 2802268
IUPAC Name 3,5,7-trimethyladamantane-1-carboxylic acid
SMILES CC12CC3(C)CC(C)(C1)CC(C2)(C3)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.33
Percent Purity 97%
Chemical Name or Material 3, 5, 7-Trimethyladamantane-1-carboxylic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.