accessibility menu, dialog, popup

UserName

CAS RN 15291-66-4

CH3OOHCH3H3C

CAS RN 15291-66-4

IUPAC Name: 3,5,7-trimethyladamantane-1-carboxylic acid
Synonyms: 3,5,7-trimethyladamantane-1-carboxylic acid
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: DHIIUPVOLLOORM-UHFFFAOYSA-N
SMILES: CC12CC3(C)CC(C)(C1)CC(C2)(C3)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results