missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5,5-Trimethylhexanoic Acid 98.0+%, TCI America™
$51.26 - $144.30
Chemical Identifiers
| CAS | 3302-10-1 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.24 |
| MDL Number | MFCD00020507 |
| InChI Key | OILUAKBAMVLXGF-UHFFFAOYNA-N |
| Synonym | hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid |
| PubChem CID | 90960 |
| IUPAC Name | 3,5,5-trimethylhexanoic acid |
| SMILES | CC(CC(=O)O)CC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
T063025ML
|
N/A | 25 mL |
Each for $51.26
|
|
|||||
|
T0630500ML
|
N/A | 500 mL |
Each for $144.30
|
|
|||||
Chemical Identifiers
| C9H18O2 | |
| MFCD00020507 | |
| hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid | |
| 3,5,5-trimethylhexanoic acid |
Specifications
| 3302-10-1 | |
| Colorless | |
| C9H18O2 | |
| hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid | |
| CC(CC(=O)O)CC(C)(C)C | |
| 158.24 | |
| 158.24 | |
| Liquid |
| -70°C | |
| 120°C | |
| MFCD00020507 | |
| OILUAKBAMVLXGF-UHFFFAOYNA-N | |
| 3,5,5-trimethylhexanoic acid | |
| 90960 | |
| ≥98.0% (T) | |
| 3,5,5-Trimethylhexanoic Acid |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : Yes