missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethylhexanoic Acid 98.0+%, TCI America™

Catalog No. T063025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T063025ML 25 mL
T0630500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T063025ML Supplier TCI America Supplier No. T063025ML

Chemical Identifiers

CAS 3302-10-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00020507
InChI Key OILUAKBAMVLXGF-UHFFFAOYNA-N
Synonym hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid
PubChem CID 90960
IUPAC Name 3,5,5-trimethylhexanoic acid
SMILES CC(CC(=O)O)CC(C)(C)C

Specifications

Melting Point -70°C
Color Colorless
Boiling Point 120°C
Quantity 25 mL
Formula Weight 158.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3,5,5-Trimethylhexanoic Acid
EINECSNumber (2)-0608
TSCA Yes