missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)acrylic Acid 97.0+%, TCI America™

Catalog No. p-7135340
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19721G 1 g
P19725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19721G Supplier TCI America Supplier No. P19721G

Chemical Identifiers

CAS 5337-79-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00040746
InChI Key SSAYTINUCCRGDR-OWOJBTEDSA-N
Synonym 3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
PubChem CID 736782
IUPAC Name (E)-3-pyridin-4-ylprop-2-enoic acid
SMILES C1=CN=CC=C1C=CC(=O)O

Specifications

Melting Point 275°C
Quantity 1 g
Formula Weight 149.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Pyridyl)acrylic Acid