accessibility menu, dialog, popup

UserName

HOON(E)

CAS RN 5337-79-1

IUPAC Name: (2E)-3-(pyridin-4-yl)prop-2-enoic acid
Synonyms: (2E)-3-(pyridin-4-yl)prop-2-enoic acid
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: SSAYTINUCCRGDR-OWOJBTEDSA-N
SMILES: OC(=O)\C=C\C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)