missing translation for 'onlineSavingsMsg'
Learn More

[3-(4-Morpholinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. ac431760500
Change view
Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431760500 50mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431760500 Supplier Thermo Scientific Chemicals Supplier No. 431760500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 480424-76-8 , 109-99-9
Molecular Formula C11H14BrMgNO
Molecular Weight (g/mol) 280.44
InChI Key VQPURMJJOOKJGN-UHFFFAOYSA-M
Synonym 3-4-morpholino methyl phenylmagnesium bromide, 3-4-morpholinylmethyl phenyl magnesiu, 4-3-bromomagnesio phenyl methyl morpholine, 3-4-morpholinylmethyl phenyl magnesium bromide solution, 3-4-morpholinylmethyl phenyl magnesium bromide solution, 0.25 m in thf, 3-4-morpholino methyl phenylmagnesium bromide 0.25 m in tetrahydrofuran, 3-4-morpholino methyl phenylmagnesium bromide, 0.25m in tetrahydrofuran, grignard reagent
PubChem CID 16217253
IUPAC Name magnesium;4-(phenylmethyl)morpholine;bromide
SMILES C1COCCN1CC2=C[C-]=CC=C2.[Mg+2].[Br-]

Specifications

CAS Min % 4.54, 89.4
CAS Max % 10.6, 95.46
Quantity 50mL
Flash Point −17°C
Solubility Information Solubility in water: reacts
Formula Weight 280.44
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.926
Density 0.9260g/mL
Chemical Name or Material [3-(4-Morpholinylmethyl)phenyl]magnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only