missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propyl isothiocyanate, 96%

Catalog No. p-7046017
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1281903 1 g
AAL1281906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1281903 Supplier Thermo Scientific Chemicals Supplier No. L1281903
Only null left

CAS: 32813-50-6 | C8H14N2OS | 186.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32813-50-6
Molecular Formula C8H14N2OS
Molecular Weight (g/mol) 186.27
MDL Number MFCD00041222
InChI Key BCEFDMYFAAAFPE-UHFFFAOYSA-N
Synonym 3-morpholinopropyl isothiocyanate, 4-3-isothiocyanatopropyl morpholine, 3-4-morpholino propyl isothiocyanate, 3-morpholinopropylisothiocyanate, 3-morpholin-4-ylpropanisothiocyanate, 3-4-morpholinyl propyl isothiocyanate, 3-morpholin-4-yl propylisothiocyanate, 4-3-isothiocyanato-propyl-morpholine, 4-3-isothiocyanatopropyl morpholine #, morpholine,4-3-isothiocyanatopropyl
PubChem CID 141745
IUPAC Name 4-(3-isothiocyanatopropyl)morpholine
SMILES S=C=NCCCN1CCOCC1

Specifications

Boiling Point 182°C to 183°C (13 mmHg)
Refractive Index 1.5305
Quantity 1 g
UN Number UN2922
Beilstein 879217
Sensitivity Moisture sensitive
Formula Weight 186.28
Percent Purity 96%
Chemical Name or Material 3-(4-Morpholinyl)propyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only