missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)phenol, 98%

CAS: 27292-49-5 | C10H13NO2 | 179.22 g/mol

Supplier:  Thermo Scientific Chemicals A1570706

 View more versions of this product

Catalog No. AAA1570706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(4-Morpholinyl)phenol
127°C to 132°C
MFCD00051675
149989
BMGSGGYIUOQZBZ-UHFFFAOYSA-N
3-morpholin-4-ylphenol
141343
98%
27292-49-5
C10H13NO2
5 g
3-morpholinophenol, 3-morpholin-4-yl phenol, phenol, 3-4-morpholinyl, 3-4-morpholinyl phenol, 3-morpholin-4-yl-phenol, n-3-hydroxyphenyl morpholine, m-morpholinophenol, 3-morpholinylphenol, 3-morpholino phenol, 3-morpholinyl-phenol
OC1=CC=CC(=C1)N1CCOCC1
179.22
179.22

Chemical Identifiers

27292-49-5
179.22
BMGSGGYIUOQZBZ-UHFFFAOYSA-N
141343
OC1=CC=CC(=C1)N1CCOCC1
C10H13NO2
MFCD00051675
3-morpholinophenol, 3-morpholin-4-yl phenol, phenol, 3-4-morpholinyl, 3-4-morpholinyl phenol, 3-morpholin-4-yl-phenol, n-3-hydroxyphenyl morpholine, m-morpholinophenol, 3-morpholinylphenol, 3-morpholino phenol, 3-morpholinyl-phenol
3-morpholin-4-ylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only