missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)phenol, 98%

Catalog No. p-7022792
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1570703 1 g
AAA1570706 5 g
AAA1570714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1570703 Supplier Thermo Scientific Chemicals Supplier No. A1570703
Only null left

CAS: 27292-49-5 | C10H13NO2 | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27292-49-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00051675
InChI Key BMGSGGYIUOQZBZ-UHFFFAOYSA-N
Synonym 3-morpholinophenol, 3-morpholin-4-yl phenol, phenol, 3-4-morpholinyl, 3-4-morpholinyl phenol, 3-morpholin-4-yl-phenol, n-3-hydroxyphenyl morpholine, m-morpholinophenol, 3-morpholinylphenol, 3-morpholino phenol, 3-morpholinyl-phenol
PubChem CID 141343
IUPAC Name 3-morpholin-4-ylphenol
SMILES OC1=CC=CC(=C1)N1CCOCC1

Specifications

Melting Point 127°C to 132°C
Quantity 1 g
Beilstein 149989
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only