missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzyl alcohol, 98%

Catalog No. AAA1266114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1266114 25 g
AAA1266122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1266114 Supplier Thermo Scientific Chemicals Supplier No. A1266114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 495-76-1 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 495-76-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00005836
InChI Key BHUIUXNAPJIDOG-UHFFFAOYSA-N
Synonym piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
PubChem CID 10322
IUPAC Name 1,3-benzodioxol-5-ylmethanol
SMILES C1OC2=C(O1)C=C(C=C2)CO

Specifications

Melting Point 50°C to 55°C
Boiling Point 156°C to 158°C (16 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 136113
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzyl alcohol
EINECSNumber 207-808-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only