missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. M26345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26345G 5 g
M26341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26345G Supplier TCI America Supplier No. M26345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 641571-11-1
Molecular Formula C11H10F3N3
Molecular Weight (g/mol) 241.217
MDL Number MFCD11846236
InChI Key WWTGXYAJVXKEKL-UHFFFAOYSA-N
Synonym 1-[3-Amino-5-(trifluoromethyl)phenyl]-4-methylimidazole
PubChem CID 12002825
IUPAC Name 3-(4-methylimidazol-1-yl)-5-(trifluoromethyl)aniline
SMILES CC1=CN(C=N1)C2=CC(=CC(=C2)C(F)(F)F)N

Specifications

Melting Point 130°C
Quantity 5 g
Formula Weight 241.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline