accessibility menu, dialog, popup

UserName

CAS RN 641571-11-1

IUPAC Name: 3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline
Synonyms: 3-(4-methylimidazol-1-yl)-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 241.22
Molecular Formula: C11H10F3N3
InChi Key: WWTGXYAJVXKEKL-UHFFFAOYSA-N
SMILES: CC1=CN(C=N1)C1=CC(N)=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results