missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 97%

Catalog No. AAH2876206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2876206 5 g
AAH2876203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2876206 Supplier Thermo Scientific Chemicals Supplier No. H2876206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 883738-27-0 | C18H29BN2O2 | 316.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 883738-27-0
Molecular Formula C18H29BN2O2
Molecular Weight (g/mol) 316.252
MDL Number MFCD09266181
InChI Key PESZAXRJFGLGTR-UHFFFAOYSA-N
Synonym 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine, 3-4-methyl-1-piperazinylmethyl benzeneboronic acid pinacol ester, 3-4-methylpiperazin-1-yl methyl phenylboronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine, amtb037, 3-4-methylpiperazin-1-yl methyl phenylboronic acid, pinacol ester, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-4-methylp ip erazine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-4-methylpiperazine
PubChem CID 46739725
IUPAC Name 1-methyl-4-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CN3CCN(CC3)C

Specifications

Quantity 5 g
Formula Weight 316.25
Percent Purity 97%
Chemical Name or Material 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only