missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)propionyl chloride, 99%

Catalog No. AAH5041606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5041606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5041606 Supplier Thermo Scientific Chemicals Supplier No. H5041606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15893-42-2 | C10H11ClO2 | 198.65 g/mol

3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with bases.

Chemical Identifiers

CAS 15893-42-2
Molecular Formula C10H11ClO2
Molecular Weight (g/mol) 198.65
MDL Number MFCD02258671
InChI Key FQVJPHCAWYRYCK-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl propionyl chloride, 3-4-methoxyphenyl propanoyl chloride, acmc-20anjm, p-methoxyhydrocinnamoyl chloride, 4-methoxyphenylpropionyl chloride, 4-methoxydihydrocinnamoyl chloride, 3-4-methoxyphenyl propanyl chloride, benzenepropanoyl chloride, 4-methoxy, 3-4-methoxyphenyl-propionyl chloride, 3-4-methoxy-phenyl-propionyl chloride
PubChem CID 10932441
IUPAC Name 3-(4-methoxyphenyl)propanoyl chloride
SMILES COC1=CC=C(CCC(Cl)=O)C=C1

Specifications

Quantity 5 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 198.65
Percent Purity 99%
Chemical Name or Material 3-(4-Methoxyphenyl)propionyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only