missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. p-7132502
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10325G 5 g
M103225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10325G Supplier TCI America Supplier No. M10325G
Only null left

Chemical Identifiers

CAS 1929-29-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00002777
InChI Key FIUFLISGGHNPSM-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy
PubChem CID 95750
IUPAC Name 3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 104°C
Boiling Point 194°C
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Methoxyphenyl)propionic Acid