missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenoxy)benzaldehyde 97.0+%, TCI America™

Catalog No. M13611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13611G Supplier TCI America Supplier No. M13611G
Only null left

Chemical Identifiers

CAS 62373-80-2
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00003358
InChI Key WLFDEVVCXPTAQA-UHFFFAOYSA-N
Synonym 3-4-methoxyphenoxy benzaldehyde, benzaldehyde, 3-4-methoxyphenoxy, acmc-209n2y, 3-4-methoxyphenoxy-benzaldehyde, 3-formyl-4'-methoxydiphenyl ether, 3-4-methoxy phenoxy benzaldehyde
PubChem CID 123553
IUPAC Name 3-(4-methoxyphenoxy)benzaldehyde
SMILES COC1=CC=C(C=C1)OC2=CC=CC(=C2)C=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 228.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(4-Methoxyphenoxy)benzaldehyde