missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxybenzoyl)acrylic acid, 98%

Catalog No. AAL1807103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1807103 1 g
AAL1807106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1807103 Supplier Thermo Scientific Chemicals Supplier No. L1807103

CAS: 5711-41-1 | C11H10O4 | 206.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5711-41-1
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.20
MDL Number MFCD00020486
InChI Key WORYXBDHTBWLLL-VOTSOKGWSA-N
Synonym 3-4-methoxybenzoyl acrylic acid, 4-4-methoxyphenyl-4-oxobut-2-enoic acid, 2e-4-4-methoxyphenyl-4-oxobut-2-enoic acid, 3-p-methoxybenzoyl acrylic acid, acrylic acid, 3-p-methoxybenzoyl, trans-3-4-methoxybenzoyl acrylic acid, e-4-4-methoxyphenyl-4-oxobut-2-enoic acid, gamma-oxo-4-methoxybenzenecrotonic acid, 4-4-methoxyphenyl-4-oxo-buten-2-saeure, 4-4-methoxyphenyl-4-oxo-2-butenoic acid
PubChem CID 5355095
IUPAC Name (E)-4-(4-methoxyphenyl)-4-oxobut-2-enoic acid
SMILES COC1=CC=C(C=C1)C(=O)\C=C\C(O)=O

Specifications

Melting Point 137°C to 140°C
Quantity 1 g
Beilstein 474529
Formula Weight 206.2
Percent Purity 98%
Chemical Name or Material 3-(4-Methoxybenzoyl)acrylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber AT2250000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only