Learn More
3,4-Lutidine, 98+%
CAS: 583-58-4 | C7H9N | 107.156 g/mol
Supplier: Thermo Scientific Chemicals L03489AP
Description
3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.
Solubility
Miscible with water, ethanol and ether.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
Specifications
| 3,4-Lutidine | |
| -11°C | |
| 163°C to 164°C | |
| Unpleasant | |
| 1.512 | |
| 500 mL | |
| 106583 | |
| Miscible with water,ethanol and ether. | |
| CC1=C(C=NC=C1)C | |
| 107.156 | |
| 107.16 |
| 583-58-4 | |
| 0.955 | |
| 47°C (116°F) | |
| C7H9N | |
| MFCD00006403 | |
| UN2929 | |
| 3,4-lutidine, pyridine, 3,4-dimethyl, unii-fcu895kj7h, pyridine,3,4-dimethyl, fcu895kj7h, 3,4-lutidene, t 3,4-lutidine, 3,4-dimethyl-pyridine, 5-20-06-00054 beilstein handbook reference | |
| NURQLCJSMXZBPC-UHFFFAOYSA-N | |
| 3,4-dimethylpyridine | |
| 11417 | |
| ≥98% |
Chemical Identifiers
| 583-58-4 | |
| 107.156 | |
| NURQLCJSMXZBPC-UHFFFAOYSA-N | |
| 3,4-dimethylpyridine |
| C7H9N | |
| MFCD00006403 | |
| 11417 | |
| CC1=C(C=NC=C1)C |
Safety and Handling
GHS H Statement
H310-H331-H226-H302-H315-H319-H335
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H332-H311-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber : 209-511-5
RTECSNumber : OK9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only