missing translation for 'onlineSavingsMsg'
Learn More

3,4-Lutidine, 98+%

Catalog No. p-7043922
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03489AC 25 mL
AAL03489AE 100 mL
AAL03489AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL03489AC Supplier Thermo Scientific Chemicals Supplier No. L03489AC

CAS: 583-58-4 | C7H9N | 107.156 g/mol

3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Lutidine is used as an intermediate in the preparation of active pharmaceutical ingredient. It is used to prepare tetrhydropyridine derivatives by reduction.

Solubility
Miscible with water, ethanol and ether.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.

Chemical Identifiers

CAS 583-58-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006403
InChI Key NURQLCJSMXZBPC-UHFFFAOYSA-N
PubChem CID 11417
IUPAC Name 3,4-dimethylpyridine
SMILES CC1=C(C=NC=C1)C

Specifications

Melting Point -11°C
Density 0.955
Boiling Point 163°C to 164°C
Flash Point 47°C (116°F)
Odor Unpleasant
Refractive Index 1.512
Quantity 25 mL
UN Number UN2929
Beilstein 106583
Synonym 3,4-lutidine, pyridine, 3,4-dimethyl, unii-fcu895kj7h, pyridine,3,4-dimethyl, fcu895kj7h, 3,4-lutidene, t 3,4-lutidine, 3,4-dimethyl-pyridine, 5-20-06-00054 beilstein handbook reference
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 107.16
Percent Purity ≥98%
Chemical Name or Material 3,4-Lutidine
Show More Show Less
Hazard Category H226-H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H310-H331-H226-H302-H315-H319-H335
Fatal in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 209-511-5
RTECSNumber OK9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only