missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Hydroxyphenyl)propionic Acid 98.0+%, TCI America™
Supplier: TCI America H0599250G
Specifications
| 3-(4-Hydroxyphenyl)propionic Acid | |
| 130°C | |
| MFCD00002778 | |
| 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid | |
| C1=CC(=CC=C1CCC(=O)O)O | |
| 166.176 | |
| CHEBI:32980 | |
| ≥98.0% (T) |
| 501-97-3 | |
| C9H10O3 | |
| 250 g | |
| NMHMNPHRMNGLLB-UHFFFAOYSA-N | |
| 3-(4-hydroxyphenyl)propanoic acid | |
| 10394 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 501-97-3 | |
| 166.176 | |
| NMHMNPHRMNGLLB-UHFFFAOYSA-N | |
| 10394 | |
| 3-(4-hydroxyphenyl)propanoic acid |
| C9H10O3 | |
| MFCD00002778 | |
| 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid | |
| CHEBI:32980 | |
| C1=CC(=CC=C1CCC(=O)O)O |
Safety and Handling
EINECSNumber : (3)-3459
RTECSNumber : MW5342000
TSCA : No