missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. p-7130630
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059925G 25 g
H0599250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H059925G Supplier TCI America Supplier No. H059925G

Chemical Identifiers

CAS 501-97-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002778
InChI Key NMHMNPHRMNGLLB-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid
PubChem CID 10394
ChEBI CHEBI:32980
IUPAC Name 3-(4-hydroxyphenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)O

Specifications

Melting Point 130°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Hydroxyphenyl)propionic Acid
EINECSNumber (3)-3459
RTECSNumber MW5342000
TSCA No