missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxy-3-methoxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. H11805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11805G 5 g
H11801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11805G Supplier TCI America Supplier No. H11805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1135-23-5
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
InChI Key BOLQJTPHPSDZHR-UHFFFAOYSA-N
Synonym 3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid
PubChem CID 14340
ChEBI CHEBI:86612
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)propanoic acid
SMILES COC1=C(C=CC(=C1)CCC(=O)O)O

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Hydroxy-3-methoxyphenyl)propionic Acid