accessibility menu, dialog, popup

UserName

H3COOHOHO

CAS RN 1135-23-5

IUPAC Name: 3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Synonyms: homovanillinic acid
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: BOLQJTPHPSDZHR-UHFFFAOYSA-N
SMILES: COC1=CC(CCC(O)=O)=CC=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)