missing translation for 'onlineSavingsMsg'
Learn More

3,4-Hexanedione, 96%

Catalog No. AC293270250
Change view
Click to view available options
Quantity:
25 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC293270250 25 mL Glass bottle
AC293272500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC293270250 Supplier Thermo Scientific Chemicals Supplier No. 293270250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4437-51-8 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4437-51-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00010237
InChI Key KVFQMAZOBTXCAZ-UHFFFAOYSA-N
Synonym 3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
PubChem CID 62539
IUPAC Name hexane-3,4-dione
SMILES CCC(=O)C(=O)CC

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -10°C
Color Yellow
Density 0.9410g/mL
Boiling Point 130°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4090 to 1.415
Linear Formula C2H5C(O)C(O)C2H5
Quantity 25 mL
Beilstein 01, 790
Specific Gravity 0.941
Solubility Information Solubility in water: 127g/L in water (20°C). Other solubilities: soluble in 1,2-propylene glycol,ethanol,oils,and fats
Formula Weight 114.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 4-Hexanedione, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-651-7
TSCA TSCA

RUO – Research Use Only