missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)propionic Acid 98.0+%, TCI America™

Catalog No. p-7129909
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08021G 1 g
F08025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08021G Supplier TCI America Supplier No. F08021G
Only null left

Chemical Identifiers

CAS 459-31-4
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00060327
InChI Key ZMKXWDPUXLPHCA-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl propionic acid, 3-4-fluorophenyl propanoic acid, benzenepropanoic acid, 4-fluoro, 3-4-fluoro-phenyl-propionic acid, 3-4-fluorophenyl propionicacid, 3-4'-fluorophenyl propanoic acid, acmc-1aehu, 4-fluorohydrocinnamic acid, 4-fluorophenylpropionic acid, 4-fluorobenzenepropanoic acid
PubChem CID 136302
IUPAC Name 3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)F

Specifications

Melting Point 89°C
Color White
Quantity 1 g
Formula Weight 168.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Fluorophenyl)propionic Acid