missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)propionic acid, 97%

Catalog No. p-7046973
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1700803 1 g
AAL1700806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1700803 Supplier Thermo Scientific Chemicals Supplier No. L1700803
Only null left

CAS: 459-31-4 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 459-31-4
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00060327
InChI Key ZMKXWDPUXLPHCA-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl propionic acid, 3-4-fluorophenyl propanoic acid, benzenepropanoic acid, 4-fluoro, 3-4-fluoro-phenyl-propionic acid, 3-4-fluorophenyl propionicacid, 3-4'-fluorophenyl propanoic acid, acmc-1aehu, 4-fluorohydrocinnamic acid, 4-fluorophenylpropionic acid, 4-fluorobenzenepropanoic acid
PubChem CID 136302
IUPAC Name 3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)F

Specifications

Melting Point 88°C to 92°C
Quantity 1 g
Beilstein 1869084
Formula Weight 168.17
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only