missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Ethylenedioxythiophene 98.0+%, TCI America™
Supplier: TCI America E074125G
Specifications
| 3,4-Ethylenedioxythiophene | |
| 112°C | |
| MFCD02093622 | |
| 2810 | |
| GKWLILHTTGWKLQ-UHFFFAOYSA-N | |
| 2,3-dihydrothieno[3,4-b][1,4]dioxine | |
| 4421864 | |
| ≥98.0% (GC) |
| 126213-50-1 | |
| C6H6O2S | |
| 25 g | |
| 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane | |
| C1COC2=CSC=C2O1 | |
| 142.172 | |
| 142.17 | |
| Liquid |
Chemical Identifiers
| 126213-50-1 | |
| 142.172 | |
| GKWLILHTTGWKLQ-UHFFFAOYSA-N | |
| 4421864 | |
| C1COC2=CSC=C2O1 |
| C6H6O2S | |
| MFCD02093622 | |
| 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane | |
| 2,3-dihydrothieno[3,4-b][1,4]dioxine |
Safety and Handling
EINECSNumber : (5)-6421
TSCA : Yes
Recommended Storage : Refrigerator