missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxythiophene 98.0+%, TCI America™

Catalog No. p-7129124
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
E07415G 5 g
E074125G 25 g
2 options

Catalog No. E07415G

Supplier: TCI America E07415G

Only null left

Chemical Identifiers

CAS 126213-50-1
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD02093622
InChI Key GKWLILHTTGWKLQ-UHFFFAOYSA-N
Synonym 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane
PubChem CID 4421864
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES C1COC2=CSC=C2O1

Specifications

Boiling Point 112°C
Quantity 5 g
UN Number 2810
Formula Weight 142.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Ethylenedioxythiophene
EINECSNumber (5)-6421
TSCA Yes
Recommended Storage Refrigerator