missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxythiophene-2,5-dicarboxylic Acid 95.0+%, TCI America™

Catalog No. E07435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07435G 5 g
E07431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07435G Supplier TCI America Supplier No. E07435G
Only null left

Chemical Identifiers

CAS 18361-03-0
Molecular Formula C8H6O6S
Molecular Weight (g/mol) 230.19
MDL Number MFCD00966477
InChI Key NWIYUAISDYJVMZ-UHFFFAOYSA-N
Synonym 2,3-Dihydrothieno[3,4-b]-1,4-dioxine-5,7-dicarboxylic Acid
PubChem CID 728841
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine-5,7-dicarboxylic acid
SMILES C1COC2=C(SC(=C2O1)C(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 230.19
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Ethylenedioxythiophene-2,5-dicarboxylic Acid