accessibility menu, dialog, popup

UserName

CAS RN 18361-03-0

OSOOHOOHO

CAS RN 18361-03-0

IUPAC Name: 2H,3H-thieno[3,4-b][1,4]dioxine-5,7-dicarboxylic acid
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxine-5,7-dicarboxylic acid
Molecular Weight (g/mol): 230.19
Molecular Formula: C8H6O6S
InChi Key: NWIYUAISDYJVMZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2OCCOC2=C(S1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results